4-(4-Amino-butyl)-piperazine-1-carboxylic acid tert-butyl ester
Catalog # APOSOR346207-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:257.372 g/mol
Storage Temperature:Ambient
CAS Number:745048-07-1
Formula:C₁₃H₂₇N₃O₂
MDL Number:MFCD04038543