Order Entry
Export
ContactUsLinkComponent
3-Methyl-isothiazole
Flammable
Irritant
3-Methyl-isothiazole
Catalog # APOSOR346143-1G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
3-Methyl-isothiazole
Catalog # APOSOR346143-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :99.154 g/mol
Boiling Pt:130…133 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:693-92-5
Formula:C₄H₅NS
MDL Number:MFCD00955579