Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(4,5-Dihydro-thiazol-2-yl)-phenol
4-(4,5-Dihydro-thiazol-2-yl)-phenol
4-(4,5-Dihydro-thiazol-2-yl)-phenol
4-(4,5-Dihydro-thiazol-2-yl)-phenol
Supplier:
Apollo Scientific
Certificates
Expand image
4-(4,5-Dihydro-thiazol-2-yl)-phenol
Supplier:
Apollo Scientific
About this item
CAS Number:
90563-68-1
:
Formula:
C₉H₉NOS
MDL Number:
MFCD00085170
:
179.239 g/mol
Storage Temperature:
Ambient