Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-2,4-dichloropyridine
3-Bromo-2,4-dichloropyridine 97%
3-Bromo-2,4-dichloropyridine 97%
APOSOR345520-5G
:
Apollo Scientific
:
3-Bromo-2,4-dichloropyridine 97%
APOSOR345520-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
226.886 g/mol
:
Ambient
:
144584-32-7
:
C₅H₂BrCl₂N
:
MFCD14705096