Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dimethoxy-2-methylbenzoic acid
4,5-Dimethoxy-2-methylbenzoic acid 97%
4,5-Dimethoxy-2-methylbenzoic acid 97%
APOSOR345046-1G
:
Apollo Scientific
:
4,5-Dimethoxy-2-methylbenzoic acid 97%
APOSOR345046-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
196.2 g/mol
:
Ambient
:
20736-28-1
:
C₁₀H₁₂O₄
:
MFCD00029889