4,5-Dichloro-2-(2-oxopropyl)-2,3-dihydropyridazin-3-one, tech
Catalog # APOSOR33382-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.041 g/mol
Storage Temperature:Ambient
CAS Number:130159-58-9
Formula:C₇H₆Cl₂N₂O₂
MDL Number:MFCD06496227