3,5-Dibromo-4,6-dimethylpyridin-2-amine, tech
Catalog # APOSOR33370-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:279.96 g/mol
Storage Temperature:Ambient
CAS Number:5407-86-3
Formula:C₇H₈Br₂N₂
MDL Number:MFCD00975063