(4E)-4-[(Dimethylamino)methylidene]-2,2-diphenyloxolan-3-one, tech
Catalog # APOSOR33319-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:293.36 g/mol
Storage Temperature:Ambient
CAS Number:339107-43-6
Formula:C₁₉H₁₉NO₂
MDL Number:MFCD00127630