Order Entry
Export
ContactUsLinkComponent
Methyl 3-(4-chlorophenyl)-4,5-dihydro-1,2-oxazole-5-carboxylate 95%
Methyl 3-(4-chlorophenyl)-4,5-dihydro-1,2-oxazole-5-carboxylate 95%
Catalog # APOSOR33248-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-(4-chlorophenyl)-4,5-dihydro-1,2-oxazole-5-carboxylate 95%
Catalog # APOSOR33248-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :239.655 g/mol
Storage Temperature:Ambient
CAS Number:91258-47-8
Formula:C₁₁H₁₀ClNO₃
MDL Number:MFCD02185946