Order Entry
Export
ContactUsLinkComponent
1-(3,4-Dimethoxyphenyl)-1H-pyrrole-2-carbaldehyde, tech
1-(3,4-Dimethoxyphenyl)-1H-pyrrole-2-carbaldehyde, tech
Catalog # APOSOR33186-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3,4-Dimethoxyphenyl)-1H-pyrrole-2-carbaldehyde, tech
Catalog # APOSOR33186-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.247 g/mol
Storage Temperature:Ambient
CAS Number:43053-78-7
Formula:C₁₃H₁₃NO₃
MDL Number:MFCD05256301