Order Entry
Export
ContactUsLinkComponent
3-Bromo-8-methylimidazo[1,2-a]pyridine 95%
Irritant
3-Bromo-8-methylimidazo[1,2-a]pyridine 95%
Catalog # APOSOR33184-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Bromo-8-methylimidazo[1,2-a]pyridine 95%
Catalog # APOSOR33184-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :211.059 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:866135-66-2
Formula:C₈H₇BrN₂
MDL Number:MFCD06496194