Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(4-Acetylphenyl)pyrrolidine-2,5-dione
1-(4-Acetylphenyl)pyrrolidine-2,5-dione, tech
1-(4-Acetylphenyl)pyrrolidine-2,5-dione, tech
APOSOR33153-1G
:
Apollo Scientific
:
1-(4-Acetylphenyl)pyrrolidine-2,5-dione, tech
APOSOR33153-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
217.221 g/mol
:
Ambient
:
36254-39-4
:
C₁₂H₁₁NO₃
:
MFCD00452866