4,6-Dimethyl-3,4-dihydro-2H-1-benzothiopyran-4-ol, tech
Catalog # APOSOR33028-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:194.293 g/mol
Storage Temperature:Ambient
CAS Number:478065-01-9
Formula:C₁₁H₁₄OS
MDL Number:MFCD02571589