Order Entry
Export
ContactUsLinkComponent
Methyl 2-(2-formyl-1H-pyrrol-1-yl)-3,3-dimethylbutanoate, tech
Methyl 2-(2-formyl-1H-pyrrol-1-yl)-3,3-dimethylbutanoate, tech
Catalog # APOSOR33000-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-(2-formyl-1H-pyrrol-1-yl)-3,3-dimethylbutanoate, tech
Catalog # APOSOR33000-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.268 g/mol
Storage Temperature:Ambient
CAS Number:343373-28-4
Formula:C₁₂H₁₇NO₃
MDL Number:MFCD01316103