Order Entry
Export
ContactUsLinkComponent
4,5-Dichloro-6-methylpyrimidin-2-amine 95%
4,5-Dichloro-6-methylpyrimidin-2-amine 95%
Catalog # APOSOR32953-1G
Supplier:  Apollo Scientific
CAS Number:  
4,5-Dichloro-6-methylpyrimidin-2-amine 95%
Catalog # APOSOR32953-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :178.019 g/mol
Storage Temperature:Ambient
CAS Number:7749-60-2
Formula:C₅H₅Cl₂N₃
MDL Number:MFCD00244693