3-{Imidazo[1,2-a]pyrimidin-2-yl}aniline, tech
Catalog # APOSOR32784-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:210.235 g/mol
Storage Temperature:Ambient
CAS Number:439108-84-6
Formula:C₁₂H₁₀N₄
MDL Number:MFCD03012714