Order Entry
Export
ContactUsLinkComponent
1-(2-Methyl-1H-indol-3-yl)-2-(piperidin-1-yl)ethane-1,2-dione, tech
1-(2-Methyl-1H-indol-3-yl)-2-(piperidin-1-yl)ethane-1,2-dione, tech
Catalog # APOSOR32665-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Methyl-1H-indol-3-yl)-2-(piperidin-1-yl)ethane-1,2-dione, tech
Catalog # APOSOR32665-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.326 g/mol
Storage Temperature:Ambient
CAS Number:330965-18-9
Formula:C₁₆H₁₈N₂O₂
MDL Number:MFCD00654545