Ethyl 2-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate 95%
Catalog # APOSOR32648-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.189 g/mol
Storage Temperature:Ambient
CAS Number:3424-43-9
Formula:C₉H₁₁NO₃
MDL Number:MFCD01871481