Order Entry
Export
ContactUsLinkComponent
3-Methyl-1-(1,4,5,6-tetrahydropyrimidin-2-yl)-4,5-dihydro-1H-pyrazol-5-one, tech
3-Methyl-1-(1,4,5,6-tetrahydropyrimidin-2-yl)-4,5-dihydro-1H-pyrazol-5-one, tech
Catalog # APOSOR32603-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Methyl-1-(1,4,5,6-tetrahydropyrimidin-2-yl)-4,5-dihydro-1H-pyrazol-5-one, tech
Catalog # APOSOR32603-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.207 g/mol
Storage Temperature:Ambient
CAS Number:338402-58-7
Formula:C₈H₁₂N₄O
MDL Number:MFCD00140900