Order Entry
Export
ContactUsLinkComponent
3,6-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid, tech
3,6-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid, tech
Catalog # APOSOR32515-1G
Supplier:  Apollo Scientific
CAS Number:  
3,6-Dichloroimidazo[1,2-a]pyridine-2-carboxylic acid, tech
Catalog # APOSOR32515-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.036 g/mol
Storage Temperature:Ambient
CAS Number:923249-11-0
Formula:C₈H₄Cl₂N₂O₂
MDL Number:MFCD08064716