4,5-Dihydro-1,3,4-benzoxadiazepin-5-one, tech
Catalog # APOSOR32497-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:162.145 g/mol
Storage Temperature:Ambient
CAS Number:144760-61-2
Formula:C₈H₆N₂O₂
MDL Number:MFCD03618004