Order Entry
Export
ContactUsLinkComponent
Methyl 2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Methyl 2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Catalog # APOSOR32412-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Catalog # APOSOR32412-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :155.151 g/mol
Storage Temperature:Ambient
CAS Number:77867-47-1
Formula:C₇H₉NO₃
MDL Number:MFCD00244305