2-Methyl-4,5-dihydro-1,3,4-benzoxadiazepin-5-one, tech
Catalog # APOSOR32350-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:176.172 g/mol
Storage Temperature:Ambient
CAS Number:144760-62-3
Formula:C₉H₈N₂O₂
MDL Number:MFCD00268745