Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(1H-1,3-Benzodiazol-2-yl)ethan-1-one
1-(1H-1,3-Benzodiazol-2-yl)ethan-1-one 95%
1-(1H-1,3-Benzodiazol-2-yl)ethan-1-one 95%
:
Apollo Scientific
1-(1H-1,3-Benzodiazol-2-yl)ethan-1-one 95%
:
Apollo Scientific
:
939-70-8
:
18773-95-0
:
C₉H₈N₂O
:
MFCD00159994
:
160.173 g/mol
:
Ambient