Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
10-Bromodecylphosphonic acid
10-Bromodecylphosphonic acid 98%
10-Bromodecylphosphonic acid 98%
Irritant
10-Bromodecylphosphonic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
10-Bromodecylphosphonic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
934985-98-5
:
Formula:
C₁₀H₂₂BrO₃P
MDL Number:
MFCD25424105
:
301.158 g/mol
:
GHS07
Storage Temperature:
Ambient