Order Entry
Export
ContactUsLinkComponent
4,7-Dichloro-1H-imidazo[4,5-d]pyridazine 95%
Irritant
4,7-Dichloro-1H-imidazo[4,5-d]pyridazine 95%
Catalog # APOSOR322895-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4,7-Dichloro-1H-imidazo[4,5-d]pyridazine 95%
Catalog # APOSOR322895-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :189.002 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:17998-43-5
Formula:C₅H₂Cl₂N₄
MDL Number:MFCD00723713