1,7-Bis-boc-1,4,7-triazaheptane
Catalog # APOSOR322863-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:303.398 g/mol
Storage Temperature:Ambient
CAS Number:117499-16-8
Formula:C₁₄H₂₉N₃O₄
MDL Number:MFCD11226825