Oxazolo[5,4-b]pyridine-2-thiol
Catalog # APOSOR322851-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:152.174 g/mol
Storage Temperature:Ambient
CAS Number:169205-99-6
:MFCD21908350
Formula:C₆H₄N₂OS
MDL Number:MFCD12755948