5-Chloro-2-hydroxybenzene-1,3-dicarbaldehyde, tech
Catalog # APOSOR32230-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.577 g/mol
Storage Temperature:Ambient
CAS Number:32596-43-3
Formula:C₈H₅ClO₃
MDL Number:MFCD00067051