Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dichloro-N-methylbenzamide
3,5-Dichloro-N-methylbenzamide 95%
3,5-Dichloro-N-methylbenzamide 95%
APOSOR32199-1G
:
Apollo Scientific
:
3,5-Dichloro-N-methylbenzamide 95%
APOSOR32199-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
204.053 g/mol
:
Ambient
:
33244-92-7
:
C₈H₇Cl₂NO
:
MFCD00243816