5-Methyl-2,4-diphenyl-2,3-dihydro-1H-pyrazol-3-one, tech
Catalog # APOSOR32181-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:250.295 g/mol
Storage Temperature:Ambient
CAS Number:79481-69-9
Formula:C₁₆H₁₄N₂O
MDL Number:MFCD00140645