Order Entry
Export
ContactUsLinkComponent
Cyclopenta-2,4-dien-1-ylidenemethyl acetate 99%
Irritant
Cyclopenta-2,4-dien-1-ylidenemethyl acetate 99%
Catalog # APOSOR321296-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Cyclopenta-2,4-dien-1-ylidenemethyl acetate 99%
Catalog # APOSOR321296-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :136.148 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:699-15-0
Formula:C₈H₈O₂
MDL Number:MFCD30749183