
(S)-4,5-Dimethyl-2-(pyrrolidin-2-yl)thiazole hydrobromide 95%
Catalog # APOSOR321282-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:263.198 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:445280-79-5
Formula:C₉H₁₄N₂S·BrH
MDL Number:MFCD30828706