Order Entry
Export
ContactUsLinkComponent
(6-Oxo-1,6-dihydropyridin-3-yl)boronic acid 97%
Toxic
(6-Oxo-1,6-dihydropyridin-3-yl)boronic acid 97%
Catalog # APOSOR321272-250MG
Supplier:  Apollo Scientific
CAS Number:  
Toxic
(6-Oxo-1,6-dihydropyridin-3-yl)boronic acid 97%
Catalog # APOSOR321272-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :138.917 g/mol
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:903899-13-8
Formula:C₅H₆BNO₃
MDL Number:MFCD22123505