Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(tert-Butyl)aniline hydrochloride
3-(tert-Butyl)aniline hydrochloride
3-(tert-Butyl)aniline hydrochloride
Irritant
3-(tert-Butyl)aniline hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-(tert-Butyl)aniline hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
83672-23-5
:
Formula:
C₁₀H₁₅N·ClH
MDL Number:
MFCD08273794
:
185.694 g/mol
Boiling Pt:
110 °C (15 mm Hg)
:
GHS07
Storage Temperature:
Ambient