Order Entry
Export
ContactUsLinkComponent
2,6-Dichloro-4-iodophenol 98%
2,6-Dichloro-4-iodophenol 98%
Catalog # APOSOR321013-250MG
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dichloro-4-iodophenol 98%
Catalog # APOSOR321013-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :288.898 g/mol
Storage Temperature:Ambient
CAS Number:34074-22-1
Formula:C₆H₃Cl₂IO
MDL Number:MFCD00466446