3-(3,3-Dimethyl-2-oxoazetidin-1-yl)benzonitrile, tech
Catalog # APOSOR32077-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:200.236 g/mol
Storage Temperature:Ambient
CAS Number:341965-81-9
Formula:C₁₂H₁₂N₂O
MDL Number:MFCD00793353