Order Entry
Export
ContactUsLinkComponent
2-Phenyl-1,2,4-triazaspiro[4.5]decan-3-one 95%
2-Phenyl-1,2,4-triazaspiro[4.5]decan-3-one 95%
Catalog # APOSOR32029-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Phenyl-1,2,4-triazaspiro[4.5]decan-3-one 95%
Catalog # APOSOR32029-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.294 g/mol
Storage Temperature:Ambient
CAS Number:73150-79-5
Formula:C₁₃H₁₇N₃O
MDL Number:MFCD00140479