Order Entry
Export
ContactUsLinkComponent
5-Methyl-3-phenyl-1,2-oxazole-4-carbonitrile 95%
5-Methyl-3-phenyl-1,2-oxazole-4-carbonitrile 95%
Catalog # APOSOR32002-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Methyl-3-phenyl-1,2-oxazole-4-carbonitrile 95%
Catalog # APOSOR32002-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :184.194 g/mol
Storage Temperature:Ambient
CAS Number:24400-67-7
Formula:C₁₁H₈N₂O
MDL Number:MFCD00172106