Order Entry
Export
ContactUsLinkComponent
2-Methyl-2,3-dihydro-1,2-benzisothiazol-3-ol 1,1-dioxide 95+%
2-Methyl-2,3-dihydro-1,2-benzisothiazol-3-ol 1,1-dioxide 95+%
Catalog # APOSOR318128-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-2,3-dihydro-1,2-benzisothiazol-3-ol 1,1-dioxide 95+%
Catalog # APOSOR318128-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :199.227 g/mol
Storage Temperature:Ambient
CAS Number:156359-48-7
Formula:C₈H₉NO₃S
MDL Number:MFCD28103339