

2-[(Trimethylsilyl)ethynyl]benzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:97308-62-8
:
Formula:C₁₂H₁₃NSi
MDL Number:MFCD09260485
:199.324 g/mol
Boiling Pt:100 °C (0.03 mm Hg)
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient