Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Hydroxy-1-methylazetidine hydrochloride
3-Hydroxy-1-methylazetidine hydrochloride
3-Hydroxy-1-methylazetidine hydrochloride
3-Hydroxy-1-methylazetidine hydrochloride
APOSOR317154-1G
:
Apollo Scientific
:
3-Hydroxy-1-methylazetidine hydrochloride
APOSOR317154-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
123.581 g/mol
:
87…95 °C
:
GHS07
:
Ambient
:
26687-49-0
:
111043-48-2
:
C₄H₉NO·ClH
:
MFCD16605978