Order Entry
Export
ContactUsLinkComponent
2-Bromo-3,5-dichlorophenol 95%
2-Bromo-3,5-dichlorophenol 95%
Catalog # APOSOR315876-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-3,5-dichlorophenol 95%
Catalog # APOSOR315876-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :241.897 g/mol
Storage Temperature:Ambient
CAS Number:13659-22-8
Formula:C₆H₃BrCl₂O
MDL Number:MFCD22414726