Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,5-Dihydroxyterephthalaldehyde

Supplier:   Apollo Scientific
2,5-Dihydroxyterephthalaldehyde
Supplier:  Apollo Scientific
Certificates
2,5-Dihydroxyterephthalaldehyde

5 Options Available Below

Product Details & Documents
  • CAS Number:1951-36-6
  • Formula:C₈H₆O₄
  • MDL Number:MFCD22055335
  • Molecular Weight:166.131 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 21,80 €
    21,80 €
    1 * 100 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 24,60 €
    24,60 €
    1 * 250 mg
       24,60 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 57,90 €
    57,90 €
    1 * 1 g
       57,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 268,00 €
    268,00 €
    1 * 5 g
       268,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 270,00 €
    1 270,00 €
    1 * 25 g
       1 270,00 €

    Specifications:
    New Product
    New Product