Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dibromo-5,6-dimethylpyridine
2,3-Dibromo-5,6-dimethylpyridine
2,3-Dibromo-5,6-dimethylpyridine
2,3-Dibromo-5,6-dimethylpyridine
APOSOR315119-1G
:
Apollo Scientific
:
2,3-Dibromo-5,6-dimethylpyridine
APOSOR315119-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
264.945 g/mol
:
Ambient
:
117846-56-7
:
C₇H₇Br₂N
:
MFCD13688994