Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichloropyrido[3,2-d]pyrimidine
4,6-Dichloropyrido[3,2-d]pyrimidine
4,6-Dichloropyrido[3,2-d]pyrimidine
Irritant
4,6-Dichloropyrido[3,2-d]pyrimidine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,6-Dichloropyrido[3,2-d]pyrimidine
Supplier:
Apollo Scientific
About this item
CAS Number:
175358-02-8
:
Formula:
C₇H₃Cl₂N₃
MDL Number:
MFCD11846204
:
200.025 g/mol
:
GHS07
Storage Temperature:
Ambient