Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,5-Dimethyl-dihydro-furan-2-one
5,5-Dimethyl-dihydro-furan-2-one
5,5-Dimethyl-dihydro-furan-2-one
Irritant
5,5-Dimethyl-dihydro-furan-2-one
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5,5-Dimethyl-dihydro-furan-2-one
Supplier:
Apollo Scientific
About this item
CAS Number:
3123-97-5
:
Formula:
C₆H₁₀O₂
MDL Number:
MFCD08061147
:
114.142 g/mol
:
GHS07
Storage Temperature:
Refrigerator