0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1,3-Diiodobicyclo[1.1.1]pentane
1,3-Diiodobicyclo[1.1.1]pentane
1,3-Diiodobicyclo[1.1.1]pentane
Irritant
1,3-Diiodobicyclo[1.1.1]pentane
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,3-Diiodobicyclo[1.1.1]pentane
Supplier:
Apollo Scientific
About this item
CAS Number:
105542-98-1
:
Formula:
C₅H₆I₂
MDL Number:
MFCD29037325
:
319.91 g/mol
:
GHS07
Storage Temperature:
Ambient