Order Entry
Export
ContactUsLinkComponent
5-Oxa-2-azaspiro[3.5]nonane oxalate 95%
Irritant
5-Oxa-2-azaspiro[3.5]nonane oxalate 95%
Catalog # APOSOR312275-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Oxa-2-azaspiro[3.5]nonane oxalate 95%
Catalog # APOSOR312275-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :217.219 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1427359-47-4
Formula:C₇H₁₃NO·C₂H₂O₄
MDL Number:MFCD20926175