Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(4,5-Dihydro-1H-imidazol-2-yl)aniline
2-(4,5-Dihydro-1H-imidazol-2-yl)aniline
2-(4,5-Dihydro-1H-imidazol-2-yl)aniline
2-(4,5-Dihydro-1H-imidazol-2-yl)aniline
Supplier:
Apollo Scientific
Certificates
Expand image
2-(4,5-Dihydro-1H-imidazol-2-yl)aniline
Supplier:
Apollo Scientific
About this item
CAS Number:
28637-61-8
:
Formula:
C₉H₁₁N₃
MDL Number:
MFCD08700379
:
161.204 g/mol
Storage Temperature:
Ambient